2,2-Difluoro-2-(4-fluoro-3-methylphenyl)acetic acid

Lieferant: Apollo Scientific

PC510322-1G
APOSPC510322-1GEA 804 EUR
APOSPC510322-1G
2,2-Difluoro-2-(4-fluoro-3-methylphenyl)acetic acid
2,2-Difluoro-2-(4-fluoro-3-methylphenyl)acetic acid
Formel: C₉H₇F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD15527112
CAS-Nummer: 1249162-93-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR