Ethyl 2,2-Difluoro-2-(4-fluoro-3-methylphenyl)acetate

Lieferant: Apollo Scientific

PC510323-1G
APOSPC510323-1GEA 804 EUR
APOSPC510323-1G
Ethyl 2,2-Difluoro-2-(4-fluoro-3-methylphenyl)acetate
Ethyl 2,2-Difluoro-2-(4-fluoro-3-methylphenyl)acetate
Formel: C₁₁H₁₁F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16036832
CAS-Nummer: 1227160-98-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR