Ethyl 2-(2-Chlorophenyl)-2,2-difluoroacetate
Lieferant: Apollo Scientific
PC510325-5G
APOSPC510325-5GEA
916
EUR
APOSPC510325-5G
Ethyl 2-(2-Chlorophenyl)-2,2-difluoroacetate
Ethyl 2-(2-Chlorophenyl)-2,2-difluoroacetate
Formel:
C₁₀H₉ClF₂O₂ Molecular Weight: 234,627 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16748873 CAS-Nummer: 1248075-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...