Ethyl 2,2-Difluoro-2-[4-(trifluoromethyl)phenyl]acetate

Lieferant: Apollo Scientific

PC510327-5G
APOSPC510327-5GEA 1010 EUR
APOSPC510327-5G
Ethyl 2,2-Difluoro-2-[4-(trifluoromethyl)phenyl]acetate
Ethyl 2,2-Difluoro-2-[4-(trifluoromethyl)phenyl]acetate
Formel: C₁₁H₉F₅O₂
Molecular Weight: 268,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17475883
CAS-Nummer: 73789-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR