2,2-Difluoro-2-(3-fluorophenyl)acetic acid

Lieferant: Apollo Scientific

PC510328-5G
APOSPC510328-5GEA 1010 EUR
APOSPC510328-5G
2,2-Difluoro-2-(3-fluorophenyl)acetic acid
2,2-Difluoro-2-(3-fluorophenyl)acetic acid
Formel: C₈H₅F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11167798
CAS-Nummer: 1042623-00-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR