6-Fluoroimidazo[1,2-a]pyridine-2-carbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
PC51033-1G
APOSPC51033-1GEA
359
EUR
APOSPC51033-1G
6-Fluoroimidazo[1,2-a]pyridine-2-carbohydrazide
6-Fluoroimidazo[1,2-a]pyridine-2-carbohydrazide
Formel:
C₈H₇FN₄O Schmelzpunkt: 263…264 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11840960 CAS-Nummer: 1135282-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...