2-(2-Cyanophenyl)-2,2-difluoroacetic acid

Lieferant: Apollo Scientific

PC510330-1G
APOSPC510330-1GEA 1010 EUR
APOSPC510330-1G
2-(2-Cyanophenyl)-2,2-difluoroacetic acid
2-(2-Cyanophenyl)-2,2-difluoroacetic acid
Formel: C₉H₅F₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD24502129
CAS-Nummer: 1824201-60-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR