Ethyl 2-(2-Cyanophenyl)-2,2-difluoroacetate

Lieferant: Apollo Scientific

PC510331-1G
APOSPC510331-1GEA 1010 EUR
APOSPC510331-1G
Ethyl 2-(2-Cyanophenyl)-2,2-difluoroacetate
Ethyl 2-(2-Cyanophenyl)-2,2-difluoroacetate
Formel: C₁₁H₉F₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29065895
CAS-Nummer: 1335290-31-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR