2-(2-Bromophenyl)-2,2-difluoroacetic acid

Lieferant: Apollo Scientific

PC510334-1G
APOSPC510334-1GEA 804 EUR
APOSPC510334-1G
2-(2-Bromophenyl)-2,2-difluoroacetic acid
2-(2-Bromophenyl)-2,2-difluoroacetic acid
Formel: C₈H₅BrF₂O₂
Molecular Weight: 251,025 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21602517
CAS-Nummer: 1375472-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR