4-(tert-Butyl)-α,α-difluorophenylacetic acid

Lieferant: Apollo Scientific

PC510338-1G
APOSPC510338-1GEA 916 EUR
APOSPC510338-1G
4-(tert-Butyl)-α,α-difluorophenylacetic acid
4-(tert-Butyl)-α,α-difluorophenylacetic acid
Formel: C₁₂H₁₄F₂O₂
Molecular Weight: 228,235 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11007715
CAS-Nummer: 1027514-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR