Ethyl 4-(tert-Butyl)-α,α-difluorophenylacetate

Lieferant: Apollo Scientific

PC510339-1G
APOSPC510339-1GEA 916 EUR
APOSPC510339-1G
Ethyl 4-(tert-Butyl)-α,α-difluorophenylacetate
Ethyl 4-(tert-Butyl)-α,α-difluorophenylacetate
Formel: C₁₄H₁₈F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11007714
CAS-Nummer: 1027514-13-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR