Ethyl 2-(3,4-Difluorophenyl)-2,2-difluoroacetate

Lieferant: Apollo Scientific

PC510340-5G
APOSPC510340-5GEA 1010 EUR
APOSPC510340-5G
Ethyl 2-(3,4-Difluorophenyl)-2,2-difluoroacetate
Ethyl 2-(3,4-Difluorophenyl)-2,2-difluoroacetate
Formel: C₁₀H₈F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11007709
CAS-Nummer: 73789-95-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR