2-(3,4-Difluorophenyl)-2,2-difluoroacetic acid

Lieferant: Apollo Scientific

PC510341-5G
APOSPC510341-5GEA 1010 EUR
APOSPC510341-5G
2-(3,4-Difluorophenyl)-2,2-difluoroacetic acid
2-(3,4-Difluorophenyl)-2,2-difluoroacetic acid
Formel: C₈H₄F₄O₂
Molecular Weight: 208,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11007708
CAS-Nummer: 915030-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR