Ethyl 2,2-Difluoro-2-(4-methoxyphenyl)acetate
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2,2-difluoro-2-(4-methoxyphenyl)acetate
PC510342-1G
APOSPC510342-1GEA
713
EUR
APOSPC510342-1G
Ethyl 2,2-Difluoro-2-(4-methoxyphenyl)acetate
Ethyl 2,2-Difluoro-2-(4-methoxyphenyl)acetate
Formel:
C₁₁H₁₂F₂O₃ Molecular Weight: 230,208 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11007701 CAS-Nummer: 112545-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...