Ethyl 2,6-Dichloro-5-fluoropyrimidine-4-carboxylate
Lieferant: Apollo Scientific
PC510346-1G
PC510346-250MG
APOSPC510346-1GEA
1120
EUR
APOSPC510346-1G
APOSPC510346-250MG
Ethyl 2,6-Dichloro-5-fluoropyrimidine-4-carboxylate
Ethyl 2,6-Dichloro-5-fluoropyrimidine-4-carboxylate
Formel:
C₇H₅Cl₂FN₂O₂ Molecular Weight: 239.031 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29065911 CAS-Nummer: 1266238-63-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...