3-(Dimethylamino)-4-fluorobenzeneboronic acid

Lieferant: Apollo Scientific

Synonyme: (3-(Dimethylamino)-4-fluorophenyl)boronic acid

PC510348-1G PC510348-5G PC510348-250MG
APOSPC510348-1GEA 384 EUR
APOSPC510348-1G APOSPC510348-5G APOSPC510348-250MG
3-(Dimethylamino)-4-fluorobenzeneboronic acid
3-(Dimethylamino)-4-fluorobenzeneboronic acid
Formel: C₈H₁₁BFNO₂
Molecular Weight: 182.988 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22573089
CAS-Nummer: 1186215-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR