6-(3-Fluorphenyl)-2-pyridincarbonsäure
Lieferant: Apollo Scientific
PC510349-1G
APOSPC510349-1GEA
493
EUR
APOSPC510349-1G
6-(3-Fluorphenyl)-2-pyridincarbonsäure
6-(3-Fluorophenyl)-2-pyridinecarboxylic acid
Formel:
C₁₂H₈FNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410053 CAS-Nummer: 887982-40-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...