Trifluoromethyl 4-methylbenzenesulphonate 95%

Lieferant: Apollo Scientific

PC510358-100MG PC510358-1G PC510358-250MG
APOSPC510358-100MGEA 0 EUR
APOSPC510358-100MG APOSPC510358-1G APOSPC510358-250MG
Trifluoromethyl 4-methylbenzenesulphonate 95%
Trifluoromethyl 4-methylbenzenesulphonate
Formel: C₈H₇F₃O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD31695793
CAS-Nummer: 175676-42-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR