4-Chloro-2-(trichloromethyl)-8-(trifluoromethyl)quinoline

Lieferant: Apollo Scientific

PC51038-1G PC51038-5G
APOSPC51038-1GEA 0 EUR
APOSPC51038-1G APOSPC51038-5G
4-Chloro-2-(trichloromethyl)-8-(trifluoromethyl)quinoline
4-Chloro-2-(trichloromethyl)-8-(trifluoromethyl)quinoline
Formel: C₁₁H₄Cl₄F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD02179716
CAS-Nummer: 91991-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR