5-(Dimethylcarbamoyl)-2-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Dimethylcarbamoyl)-2-fluorobenzeneboronic acid, (5-(Dimethylcarbamoyl)-2-fluorophenyl)boronic acid, 5-Dimethylcarbamoyl-2-fluorobenzeneboronic acid

PC5104-1G
APOSPC5104-1GEA 176 EUR
APOSPC5104-1G
5-(Dimethylcarbamoyl)-2-fluorphenylboronsäure 98%
5-(Dimethylcarbamoyl)-2-fluorphenylboronsäure
Formel: C₉H₁₁BFNO₃
Molecular Weight: 211,00 g/mol
Schmelzpunkt: 143…147 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235102
CAS-Nummer: 874289-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR