4-Chlor-2-methyl-7-(trifluormethyl)chinolin

Lieferant: Apollo Scientific

PC51040-1G
APOSPC51040-1GEA 353 EUR
APOSPC51040-1G
4-Chlor-2-methyl-7-(trifluormethyl)chinolin
4-Chloro-2-methyl-7-(trifluoromethyl)quinoline
Formel: C₁₁H₇ClF₃N
Molecular Weight: 245.63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272334
CAS-Nummer: 18529-09-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR