4-Chloro-5,7-difluoroquinoline
Lieferant: Apollo Scientific
|
Achtung
|
PC51041-1G
APOSPC51041-1GEA
168
EUR
APOSPC51041-1G
4-Chloro-5,7-difluoroquinoline
4-Chloro-5,7-difluoroquinoline
Formel:
C₉H₄ClF₂N Molecular Weight: 199,585 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06808522 CAS-Nummer: 874831-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...