4-Chloro-5,8-difluoroquinoline

Lieferant: Apollo Scientific

PC51042-1G
APOSPC51042-1GEA 161 EUR
APOSPC51042-1G
4-Chloro-5,8-difluoroquinoline
4-Chloro-5,8-difluoroquinoline
Formel: C₉H₄ClF₂N
Molecular Weight: 199,585 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407958
CAS-Nummer: 1208626-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR