4-Chloro-2-(trichloromethyl)-6-(trifluoromethyl)quinoline

Lieferant: Apollo Scientific

PC51044-1G
APOSPC51044-1GEA 351 EUR
APOSPC51044-1G
4-Chloro-2-(trichloromethyl)-6-(trifluoromethyl)quinoline
4-Chloro-2-(trichloromethyl)-6-(trifluoromethyl)quinoline
Formel: C₁₁H₄Cl₄F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD00272420
CAS-Nummer: 91991-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR