2-Fluor-5-(phenylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: (2-Fluoro-5-(phenylcarbamoyl)phenyl)boronic acid

PC5105-1G
APOSPC5105-1GEA 248 EUR
APOSPC5105-1G
2-Fluor-5-(phenylcarbamoyl)phenylboronsäure 98%
2-Fluor-5-(phenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BFNO₃
Schmelzpunkt: 252…256 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235095
CAS-Nummer: 874289-41-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR