6-(Trifluormethyl)-1H-indazol 95%

Lieferant: Apollo Scientific
Achtung

PC51050-1G
APOSPC51050-1GEA 105 EUR
APOSPC51050-1G
6-(Trifluormethyl)-1H-indazol 95%
6-(Trifluoromethyl)-1H-indazole
Formel: C₈H₅F₃N₂
Molecular Weight: 186.1300 g/mol
Schmelzpunkt: 119…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08690097
CAS-Nummer: 954239-22-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR