2-Nitro-3,4,5-trifluoroaniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Amino-2,3,4-trifluoronitrobenzene

PC51051-1G
APOSPC51051-1GEA 87.3 EUR
APOSPC51051-1G
2-Nitro-3,4,5-trifluoroaniline
2-Nitro-3,4,5-trifluoroaniline
Formel: C₆H₃F₃N₂O₂
Molecular Weight: 192,095 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20036244
CAS-Nummer: 882068-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR