5-(Cyclohexylcarbamoyl)-2-fluorobenzeneboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(5-(Cyclohexylcarbamoyl)-2-fluorophenyl)boronic acid, 5-(Cyclohexylcarbmoyl)-2-fluorobenzeneboronic acid
PC5106-1G
APOSPC5106-1GEA
154
EUR
APOSPC5106-1G
5-(Cyclohexylcarbamoyl)-2-fluorobenzeneboronic acid 98%
5-(Cyclohexylcarbamoyl)-2-fluorobenzeneboronic acid
Formel:
C₁₃H₁₇BFNO₃ Molecular Weight: 265,088 g/mol Schmelzpunkt: 164 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235096 CAS-Nummer: 874289-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...