(1H,1H,2H,2H-Tridecafluorooct-1-yl)phosphonic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooct-1-yl)phosphonic acid, [2-(Perfluorohex-1-yl)ethyl]phosphonic acid

PC51063-1G PC51063-5G
APOSPC51063-1GEA 122 EUR
APOSPC51063-1G APOSPC51063-5G
(1H,1H,2H,2H-Tridecafluorooct-1-yl)phosphonic acid
(1H,1H,2H,2H-Tridecafluorooct-1-yl)phosphonic acid
Formel: C₈H₆F₁₃O₃P
Schmelzpunkt: 168…173 °C
Lagertemperatur: Kühlschrank
MDL: MFCD18910708
CAS-Nummer: 252237-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR