Perfluoro(1-chlorocyclopent-1-ene)
Lieferant: Apollo Scientific
PC51068-1G
PC51068-250MG
APOSPC51068-1GEA
116
EUR
APOSPC51068-1G
APOSPC51068-250MG
Perfluoro(1-chlorocyclopent-1-ene)
Perfluoro(1-chlorocyclopent-1-ene)
Formel:
C₅ClF₇ Schmelzpunkt: 56…58 °C Dichte: 1,606 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD19442032 CAS-Nummer: 1759-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...