4-Fluor-3-(methoxycarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: 4-Fluoro-3-(methoxycarbamoyl)benzeneboronic acid

PC5107-1G
APOSPC5107-1GEA 176 EUR
APOSPC5107-1G
4-Fluor-3-(methoxycarbamoyl)phenylboronsäure 98%
4-Fluor-3-(methoxycarbamoyl)phenylboronsäure
Formel: C₈H₉BFNO₄
Molecular Weight: 212,97 g/mol
Schmelzpunkt: 213…215 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235105
CAS-Nummer: 913835-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR