4-Amino-3-fluorobenzeneboronic acid, N-CBZ protected 98%

Lieferant: Apollo Scientific

Synonyme: 4-(Benzyloxycarbonylamino)-3-fluorobenzeneboronic acid

PC5108-1G PC5108-5G
APOSPC5108-1GEA 139 EUR
APOSPC5108-1G APOSPC5108-5G
4-Amino-3-fluorobenzeneboronic acid, N-CBZ protected 98%
4-Amino-3-fluorobenzeneboronic acid, N-CBZ protected
Formel: C₁₄H₁₃BFNO₄
Molecular Weight: 289.067 g/mol
Schmelzpunkt: 197…199 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235098
CAS-Nummer: 874290-60-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR