3-Fluorophenylsulphur pentafluoride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Fluoro-3-(pentafluorothio)benzene, 1-Fluoro-3-(pentafluorosulphanyl)benzene
PC51096-1G
PC51096-5G
APOSPC51096-1GEA
145
EUR
APOSPC51096-1G
APOSPC51096-5G
3-Fluorophenylsulphur pentafluoride
3-Fluorophenylsulphur pentafluoride
Formel:
C₆H₄F₆S Molecular Weight: 222.151 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21362225 CAS-Nummer: 1422-41-9 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...