2,4-Difluoro-5-iodonitrobenzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,5-Difluoro-2-iodo-4-nitrobenzene
PC51101-1G
PC51101-250MG
PC51101-5G
PC51101-25G
APOSPC51101-1GEA
21.9
EUR
APOSPC51101-1G
APOSPC51101-250MG
APOSPC51101-5G
APOSPC51101-25G
2,4-Difluoro-5-iodonitrobenzene
2,4-Difluoro-5-iodonitrobenzene
Formel:
C₆H₂F₂INO₂ Siedepunkt: 65…67 °C (0,1 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD20322039 CAS-Nummer: 148388-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...