2-Bromo-3-iodobenzotrifluoride 97%

Lieferant: Apollo Scientific
Achtung

PC51105-100MG PC51105-1G PC51105-250MG
APOSPC51105-100MGEA 143 EUR
APOSPC51105-100MG APOSPC51105-1G APOSPC51105-250MG
2-Bromo-3-iodobenzotrifluoride 97%
2-Bromo-3-iodobenzotrifluoride
Formel: C₇H₃BrF₃I
Lagertemperatur: Kühlschrank
MDL: MFCD17012838
CAS-Nummer: 1049731-01-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR