3,5-Bis(trifluoromethyl)benzyl trifluoromethyl sulphone 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Bis(trifluoromethyl)-5-{[(trifluoromethyl)sulphonyl]methyl}benzene

PC51108-1G
APOSPC51108-1GEA 340 EUR
APOSPC51108-1G
3,5-Bis(trifluoromethyl)benzyl trifluoromethyl sulphone 97%
3,5-Bis(trifluoromethyl)benzyl trifluoromethyl sulphone
Formel: C₁₀H₅F₉O₂S
Molecular Weight: 360,196 g/mol
Schmelzpunkt: 85…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD20489423
CAS-Nummer: 434957-15-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR