2-(Trifluoromethoxy)benzyl trifluoromethyl sulphone

Lieferant: Apollo Scientific
Achtung

Synonyme: α,α,α-Trifluoro-2-{[(trifluoromethyl)sulphonyl]methyl}anisole, 1-(Trifluoromethoxy)-2-{[(trifluoromethyl)sulphonyl]methyl}benzene

PC51111-1G
APOSPC51111-1GEA 340 EUR
APOSPC51111-1G
2-(Trifluoromethoxy)benzyl trifluoromethyl sulphone
2-(Trifluoromethoxy)benzyl trifluoromethyl sulphone
Formel: C₉H₆F₆O₃S
Molecular Weight: 308,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20489422
CAS-Nummer: 1346521-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR