5-Bromo-2-fluoro-3-hydroxypyridine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-3-hydroxy-5-bromopyridine, 5-Bromo-2-fluoropyridin-3-ol
PC51114-5G
APOSPC51114-5GEA
94.4
EUR
APOSPC51114-5G
5-Bromo-2-fluoro-3-hydroxypyridine 98%
5-Bromo-2-fluoro-3-hydroxypyridine
Formel:
C₅H₃BrFNO Molecular Weight: 191.986 g/mol Schmelzpunkt: 146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11040265 CAS-Nummer: 1012084-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...