3-Bromo-5-fluoro-4-hydroxypyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-5-fluoropyridin-4-ol

PC51116-1G
APOSPC51116-1GEA 509 EUR
APOSPC51116-1G
3-Bromo-5-fluoro-4-hydroxypyridine
3-Bromo-5-fluoro-4-hydroxypyridine
Formel: C₅H₃BrFNO
Molecular Weight: 191,986 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185370
CAS-Nummer: 1214342-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR