3-Hydroxy-2-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Trifluoromethyl)pyridin-3-ol, 3-Hydroxy-α,α,α-trifluoro-2-picoline
PC51117-1G
APOSPC51117-1GEA
328
EUR
APOSPC51117-1G
3-Hydroxy-2-(trifluoromethyl)pyridine
3-Hydroxy-2-(trifluoromethyl)pyridine
Formel:
C₆H₄F₃NO Schmelzpunkt: 135…137 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185667 CAS-Nummer: 1063697-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...