2-Bromo-5-fluoro-4-hydroxypyridine

Lieferant: Apollo Scientific
Achtung

PC51118-1G PC51118-250MG
APOSPC51118-1GEA 321 EUR
APOSPC51118-1G APOSPC51118-250MG
2-Bromo-5-fluoro-4-hydroxypyridine
2-Bromo-5-fluoro-4-hydroxypyridine
Formel: C₅H₃BrFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD13189884
CAS-Nummer: 1196152-88-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR