2-Bromo-3-fluoro-4-hydroxypyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-3-fluoropyridin-4-ol

PC51120-250MG
APOSPC51120-250MGEA 21.9 EUR
APOSPC51120-250MG
2-Bromo-3-fluoro-4-hydroxypyridine
2-Bromo-3-fluoro-4-hydroxypyridine
Formel: C₅H₃BrFNO
Molecular Weight: 191,986 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18257612
CAS-Nummer: 1211525-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR