3-Fluoro-4-hydroxypyridine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 3-Fluoropyridin-4-ol hydrochloride

PC51122-1G
APOSPC51122-1GEA 190 EUR
APOSPC51122-1G
3-Fluoro-4-hydroxypyridine hydrochloride
3-Fluoro-4-hydroxypyridine hydrochloride
Formel: C₅H₄FNO·ClH
Molecular Weight: 149,551 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19442154
CAS-Nummer: 1309602-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR