2-Fluoro-5-hydroxy-4-methylpyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Fluoro-4-methylpyridin-3-ol, 2-Fluoro-5-hydroxy-4-picoline

PC51126-1G
APOSPC51126-1GEA 161 EUR
APOSPC51126-1G
2-Fluoro-5-hydroxy-4-methylpyridine
2-Fluoro-5-hydroxy-4-methylpyridine
Formel: C₆H₆FNO
Molecular Weight: 127,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16611074
CAS-Nummer: 1227577-18-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR