5-Fluoro-4-hydroxy-2-methoxypyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-2-methoxypyridin-4(1H)-one, 5-Fluoro-4-hydroxypyridin-2-yl methyl ether, 5-Fluoro-2-methoxypyridin-4-ol

PC51127-1G
APOSPC51127-1GEA 575 EUR
APOSPC51127-1G
5-Fluoro-4-hydroxy-2-methoxypyridine
5-Fluoro-4-hydroxy-2-methoxypyridine
Formel: C₆H₆FNO₂
Molecular Weight: 143,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189883
CAS-Nummer: 51173-14-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR