5-Bromo-2-chloro-4-fluorophenylacetonitrile 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(5-Bromo-2-chloro-4-fluorophenyl)acetonitrile, 5-Bromo-2-chloro-4-fluorobenzyl cyanide

PC51137-25G
APOSPC51137-25GEA 148 EUR
APOSPC51137-25G
5-Bromo-2-chloro-4-fluorophenylacetonitrile 97%
5-Bromo-2-chloro-4-fluorophenylacetonitrile
Formel: C₈H₄BrClFN
Molecular Weight: 248.479 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23166759
CAS-Nummer: 1426290-08-5
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR