2-Bromo-4-(trifluoromethoxy)phenylacetylene

Lieferant: Apollo Scientific
Achtung

PC51142-1G PC51142-250MG
APOSPC51142-1GEA 145 EUR
APOSPC51142-1G APOSPC51142-250MG
2-Bromo-4-(trifluoromethoxy)phenylacetylene
2-Bromo-4-(trifluoromethoxy)phenylacetylene
Formel: C₉H₄BrF₃O
Lagertemperatur: Kühlschrank
MDL: MFCD23166760
CAS-Nummer: 1426290-11-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR