3,5-Bis(trifluoromethyl)phenyl propyl ether

Lieferant: Apollo Scientific

Synonyme: α,α,α,α',α',α'-Hexafluoro-5-propoxy-m-xylene, 1,3-Bis(trifluoromethyl)-5-propoxybenzene

PC51145-1G
APOSPC51145-1GEA 60.9 EUR
APOSPC51145-1G
3,5-Bis(trifluoromethyl)phenyl propyl ether
3,5-Bis(trifluoromethyl)phenyl propyl ether
Formel: C₁₁H₁₀F₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD28396341
CAS-Nummer: 1420977-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR