2-Fluoro-6-methylbenzyl alcohol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(6-Fluoro-o-tolyl)methanol, (2-Fluoro-6-methylphenyl)methanol
PC51146-1G
APOSPC51146-1GEA
56.7
EUR
APOSPC51146-1G
2-Fluoro-6-methylbenzyl alcohol 97%
2-Fluoro-6-methylbenzyl alcohol
Formel:
C₈H₉FO Molecular Weight: 140.155 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660280 CAS-Nummer: 478163-35-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...