3-(Pentafluorothio)phenylacetylene 96%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Ethynyl-3-(pentafluorosulphanyl)benzene, 3-Ethynylphenylsulphur pentafluoride
PC51148-1G
PC51148-250MG
PC51148-5G
APOSPC51148-1GEA
422
EUR
APOSPC51148-1G
APOSPC51148-250MG
APOSPC51148-5G
3-(Pentafluorothio)phenylacetylene 96%
3-(Pentafluorothio)phenylacetylene
Formel:
C₈H₅F₅S Molecular Weight: 228.182 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD23166761 CAS-Nummer: 1426290-10-9 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...